Mouse Recombinant Glioma culture PB (from E. coli)
Catalog # 77753-630
Supplier: VWR International
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:Each
- Storage conditions:At or below –20 °C
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:Glioma culture PB
- Endotoxin level:Low
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
About this item
Cytokines for cell culture of mouse glioma cells. A glioma is a generic term used to describe tumors that arise from the glia tissue of the brain that supports and surrounds neurons. Glioma tumor cells can be successfully cultured ex vivo with EGF, FGF-basic (or FGF-2), and PDGFs (AA and BB).
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.