Dog Recombinant IL-2 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 2 (IL-2) is an immunomodulatory cytokine that is produced by lymphocytes. IL-2 signals through the IL-2R receptor to induce activated T cell proliferation and promote T cell differentiation. IL-2 also stimulates the proliferation and differentiation of B cells, natural killer cells, monocytes, and macrophages.
- High quality Low endotoxin Lot-to-lot consistency Animal free Manufactured following GMP guidelines
Caution: For research use or further manufacturing only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Dog
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: CTLL-2 cell proliferation; ED50 ≤5 ng/ml; Bioactivity ≥2.0×10⁵ units/mg
- Gene ID:Q29416
- Reconstitution instructions:Sterile 10 mM sodium bicarbonate, pH 8.5 at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:T cell growth factor|TCGF|Aldesleukin
- Protein/peptide name:IL-2
- Purity:≥95%
- Molecular weight:15.6
- Sequence:MAPITSSSTK ETEQQMEQLL LDLQLLLNGV NNYENPQLSR MLTFKFYTPK KATEFTHLQC LAEELKNLEE VLGLPQSKNV HLTDTKELIS NMNVTLLKLK GSETSYNCEY DDETATITEF LNKWITFSQS IFSTLT
- Endotoxin level:Low
- Formulation:20 mM Sodium bicarbonate, pH 8.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together