Mouse Recombinant IL-5 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 5 (IL-5) is a hematopoietic growth factor that is expressed in type 2 T helper (Th2) cells, mast cells, and eosinophils. IL-5 acts through the IL-5 receptor (IL-5R), stimulates B cell growth, and mediates eosinophil activation. IL-5 expression is regulated by the GATA binding protein 3 (GATA3) transcription factor. Human and mouse IL-5 show cross-reactivity.
- High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: TF-1 cell proliferation; ED50 ≤2 ng/ml; Bioactivity ≥5.0×10⁵ units/mg
- Gene ID:P04401
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:EDF|BCDFII|TRF
- Protein/peptide name:IL-5
- Purity:≥95%
- Molecular weight:13.1/26.3
- Sequence:MEIPMSTVVK ETLTQLSAHR ALLTSNETMR LPVPTHKNHQ LCIGEIFQGL DILKNQTVRG GTVEMLFQNL SLIKKYIDRQ KEKCGEERRR TRQFLDYLQE FLGVMSTEWA MEG
- Endotoxin level:Low
- Formulation:20 mM Sodium bicarbonate, pH 9.0
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together