Human Recombinant IL-9 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 9 (IL-9) is a cytokine produced by type 2 T helper (Th2) cells and regulates hematopoietic cells. IL-9 signals through the interleukin 9 receptor (IL9R) to activate STAT signaling. IL-9 functions to induce cell proliferation, prevent cell apoptosis, and is associated with asthma and airway hyperresponsiveness.
- High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: MO7e cell proliferation; ED50 ≤2 ng/ml; Bioactivity ≥5.0×10⁵ units/mg
- Gene ID:P15248
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:T-cell growth factor p40|P40 cytokine
- Protein/peptide name:IL-9
- Purity:≥95%
- Molecular weight:14.3
- Sequence:MQGCPTLAGI LDINFLINKM QEDPASKCHC SANVTSCLCL GIPSDNCTRP CFSERLSQMT NTTMQTRYPL IFSRVKKSVE VLKNNKCPYF SCEQPCNQTT AGNALTFLKS LLEIFQKEKM RGMRGKI
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together