Human Recombinant IL-1B (from E. coli)
New Product
Interleukin 1 beta (IL-1β) is a pro-inflammatory cytokine that is produced by monocytes, macrophages, and dendritic cells (DCs). IL-1β signals through the IL-1 receptor, type 1 (IL-1R1) to activate the myeloid differentiation primary response 88 (MYD88) signaling pathway, which contains the cytoplasmic Toll/IL-1 receptor (TIR) domain adapter. IL-1β and the independently regulated IL-1α protein have overlapping pro-inflammatory activities to induce adhesion molecule expression on epithelial cells, control fever induction, initiate rheumatoid arthritis, and promote septic shock.
- High quality
- Low endotoxin
- Affordable
:
For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: D10S cell proliferation; ED50 ≤0.012 ng/ml; Bioactivity ≥8.3×10⁷ units/mg
- Gene ID:P01584
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Catabolin|LAF|EP|LEM|MCF
- Protein/peptide name:IL-1B
- Purity:≥95%
- Molecular weight:17.5
- Sequence:MAPVRSLNCT LRDSQQKSLV MSGPYELKAL HLQGQDMEQQ VVFSMSFVQG EESNDKIPVA LGLKEKNLYL SCVLKDDKPT LQLESVDPKN YPKKKMEKRF VFNKIEINNK LEFESAQFPN WYISTSQAEN MPVFLGGTKG GQDITDFTMQ FVSS
- Endotoxin level:Low
- Formulation:10 mM Sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Frequently Bought Together