Human Recombinant Rantes (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Regulated upon activation, normal T cell expressed and secreted (RANTES), also called CCL5, is a chemokine produced by T cells three to five days after T cell activation. Rantes signals through G protein-coupled receptors CCR5, CCR3, CCR1, and through the human CMV-encoded viral receptor US28. Rantes functions to recruit immune cells to inflammatory sites.
High quality
- Low endotoxin
- Affordable
Caution:
For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: THP-1 chemotaxis; ED₅₀ ≤250 ng/ml; Bioactivity ≥4.0×10³ units/mg
- Gene ID:P13501
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CCL5
- Protein/peptide name:Rantes
- Purity:≥95%
- Molecular weight:7.9 kDa
- Sequence:SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together