Order Entry
Canada
ContactUsLinkComponent
COVID-19 Sequencing Kit
COVID-19 Sequencing Kit
Catalog # 77867-954
New Product
CAS Number:  
undefined
COVID-19 Sequencing Kit
Catalog # 77867-954
Supplier Number:  C19MINI114PK
New Product
CAS Number:  
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Description:
    COVID mini 576 samples
  • Size:
    1 pack
  • Environmentally Preferable:
  • Compatible with:
    R10.4.1 Flow Cells
  • No. of Reactions:
    576

Specifications

About this item

A simple, scalable and rapid method for sequencing up to 576 SARS-CoV-2 samples.  Contains all Oxford Nanopore sequencing reagents for rapid whole-genome sequencing of SARS-CoV-2.

  • Midnight Expansion Kit with RT-PCR reagents and primers to amplify SARS-CoV-2 from input RNA

  • Rapid Barcoding Kits 96 V14 to barcode and prepare the amplified material for sequencing
  • MinION/GridION Flow Cells to perform sequencing on MinION or GridION devices

The COVID Mini V14 bundle contains: 
- 1× Midnight Expansion Kit (EXP-MRT001)
- 2× Rapid Barcoding Kit 96 V14 (SQK-RBK114.96)
- 6× MinION/GridION Flow Cells (FLO-MIN114)

The Midnight method amplifies the SARS-CoV-2 genome in 29 overlapping amplicons (~1,200 bp long). By amplifying longer amplicons, Oxford Nanopore's rapid chemistry can be deployed, enabling users to rapidly attach a barcode, pool samples and adapt them for sequencing. This method is simple and quick to deploy. It allows rapid turnaround times that can assist with monitoring of novel variants of concern.

The protocol delivers robust full-genome coverage across a wide range of Ct values.

Users can pair the library preparation with an Oxford Nanopore analysis solution to enable a complete end-to-end workflow. The workflow and bundle are designed to run 96 samples at a time. Users wishing to run in multiples of 24 will need to buy 3× Flow Cell Wash Kits (EXP-WSH004).

Sample multiplexing: ≤96

Delivery:

Flow cells and kits are shipped together at 2 to 8 °C. The packaging is designed to protect the flow cells from freezing. R10.4.1 Flow Cells can be stored unopened at 2 to 8 °C for 12 weeks or unopened at room temperature for one month. Kits should be stored at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.

Caution:

Not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.