Human/Mouse Recombinant H/M TGF-B 3 (from E. coli cells)
New Product
Transforming growth factors (TGFs) are multifunctional peptides that regulate growth and differentiation in most cell types. The TGF-β family of proteins signal through serine/threonine kinase receptors. TGF-β isoforms (TGF-β1, -β2, and –β3) have overlapping, yet distinct biological actions in developing and adult tissues. TGF-β3 is an important factor in regulating cell adhesion and accelerating wound repair. TGF-β3 also functions during osteoblast proliferation, chemotaxis, and collagen synthesis.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:HumanMouse
- Storage conditions:Store at 4 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation; ED50 ≤1 ng/ml; Bioactivity ≥1.0×10⁶ units/mg
- Gene ID:P10600/P17125
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein/peptide name:H/M TGF-B 3
- Purity:≥95%
- Molecular weight:12.9/25.7 kDa
- Sequence:MALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCS
- Endotoxin level:Low
- Formulation:10 mM acetic acid and 20% ethanol at a concentration of 0.25 mg/ml
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:Ice pack
Frequently Bought Together