About this item
GM-CSF is a hematopoietic growth factor with the ability to form colonies of granulocytes and macrophages. Enhances the number of circulating white blood cells and function of neutrophils and monocytes. Regulates multiple functions via transcription factors in the differentiated cells, including cell survival, proliferation, and maturation. Generates in vitro murine and human DC populations from bone marrow precursors and blood monocytes, respectively.
- High quality
- Low endotoxin
- Lot-to-lot consistency
- Animal free
- Manufactured following GMP guidelines
Human and mouse GM-CSF do not show cross-reactivity. The biological activity of CTGrade GMP interleukins and growth factors is standardized, where applicable, to WHO International standards, providing cell and gene therapy developers consistent, lot-to-lot biological activity and performance.
Host cell protein: ≤30 ng/ml, host cell DNA: ≤0.01 ng/ml, sterility testing: No growth, mycoplasma: Negative
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:At or below –20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.05
- Biological activity:Bioactivity assay: Proliferation of TF-1; ED50 ≤0.2 ng/ml; Bioactivity ≥1.6×10⁴ IU/µg
- Gene ID:P04141
- Reconstitution instructions:Sterile water at 0.1 - 0.5 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:CSF-2|Pluripoietin-α|MGI-1GM
- Protein/peptide name:GMCSF
- Purity:≥98%
- Molecular weight:14.6 kDa
- Sequence:MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:GMP
- Shipping temperature:Ambient