Mouse Recombinant RESISTIN (from E. coli cells)
New Product
Resistin, also known as FIZZ3, is a peptide hormone belonging to a class of cysteine-rich secreted proteins termed the resistin-like molecules (RELM) family. Mouse resistin, produced by adipocytes, is involved in insulin resistance and modulates glucose homeostasis and adipogenesis.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q5BMX4
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:FIZZ3|ADSF
- Protein/peptide name:RESISTIN
- Purity:≥95%
- Molecular weight:10.3/20.6 kDa
- Sequence:MSSMPLCPID EAIDKKIKQD FNSLFPNAIK NIGLNCWTVS SRGKLASCPE GTAVLSCSCG SACGSWDIRE EKVCHCQCAR IDWTAARCCK LQVAS
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Frequently Bought Together