Mouse Recombinant LEPTIN (from E. coli cells)
Supplier: VWR International
New Product
Certificates
About this item
Leptin is a hormone that is produced by adipose tissue and plays critical roles in the physiologic regulation of body weight. Leptin acts through the leptin receptor (LEPR) to regulate adipose mass by inhibiting hunger and balancing energy usage.
- High quality
- Low endotoxin
- Affordable
Leptin mutations cause severe hereditary obesity and hypogonadism in rodents and humans. Leptin also has thermogenic actions, regulates enzymes of fatty acid oxidation, and is involved in hematopoiesis, angiogenesis, wound healing, inflammation, and immune responses.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q544U0
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Obesity protein
- Protein/peptide name:LEPTIN
- Purity:≥95%
- Molecular weight:16.1 kDa
- Sequence:MVPIQKVQDD TKTLIKTIVT RINDISHTQS VSAKQRVTGL DFIPGLHPIL SLSKMDQTLA VYQQVLTSLP SQNVLQIAND LENLRDLLHL LAFSKSCSLP QTSGLQKPES LDGVLEASLY STEVVALSRL QGSLQDILQQ LDVSPEC
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together