Human Recombinant PSP (from E. coli cells)
Supplier: VWR International
New Product
Certificates
About this item
Persephin is a neurotrophic factor of the glial cell line-derived neurotrophic factor (GDNF) family. Persephin promotes survival and growth of dopaminergic and motor neurons, but not peripheral neurons. Persephin is a ligand for the RET receptor tyrosine kinase.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: TT cell proliferation; ED50 ≤20 ng/ml; Bioactivity ≥5.0×10⁴ units/mg
- Gene ID:O60542
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:PSP
- Protein/peptide name:PSP
- Purity:≥95%
- Molecular weight:10.4/20.8 kDa
- Sequence:MALSGPCQLW SLTLSVAELG LGYASEEKVI FRYCAGSCPR GARTQHGLAL ARLQGQGRAH GGPCCRPTRY TDVAFLDDRH RWQRLPQLSA AACGCGG
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together