Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant PSP (from E. coli cells)
New Product
 
undefined
Human Recombinant PSP (from E. coli cells)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: TT cell proliferation; ED50 ≤20 ng/ml; Bioactivity ≥5.0×10⁴ units/mg
  • Gene ID:
    O60542
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    PSP
  • Protein/peptide name:
    PSP
  • Purity:
    ≥95%
  • Molecular weight:
    10.4/20.8 kDa
  • Sequence:
    MALSGPCQLW SLTLSVAELG LGYASEEKVI FRYCAGSCPR GARTQHGLAL ARLQGQGRAH GGPCCRPTRY TDVAFLDDRH RWQRLPQLSA AACGCGG
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

 

Frequently Bought Together