Order Entry
Canada
ContactUsLinkComponent
Human Recombinant FGF-ACIDIC (from E. coli cells)
Human Recombinant FGF-ACIDIC (from E. coli cells)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant FGF-ACIDIC (from E. coli cells)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: NR6R-3T3 cell proliferation; ED50 ≤2 ng/ml; Bioactivity ≥5.0×10⁵ units/mg
  • Gene ID:
    P05230
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    FGF1|HBGF-1|Beta-endothelial growth factor|ECGF-beta
  • Protein/peptide name:
    FGF-ACIDIC
  • Purity:
    ≥95%
  • Molecular weight:
    16 kDa
  • Sequence:
    MFNLPPGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SVGEVYIKST ETGQYLAMDT DGLLYGSQTP NEECLFLERL EENHYNTYIS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 150 mM sodium sulfate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together