Human Recombinant FGF-ACIDIC (from E. coli cells)
Supplier: VWR International
New Product
Certificates
About this item
Acidic fibroblast growth factor (FGF-acidic), also known as FGF-1, is a potent inducer of DNA synthesis, cell proliferation, and has chemotactic activities. FGF-acidic regulates cardiogenesis through protein kinase C signaling. FGF-acidic also functions as an insulin sensitizer and mediates adipose tissue remodeling. High serum levels of FGF-acidic are associated with type 2 diabetes mellitus (T2DM), suggesting a pathogenic role of FGF-acidic during T2DM.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: NR6R-3T3 cell proliferation; ED50 ≤2 ng/ml; Bioactivity ≥5.0×10⁵ units/mg
- Gene ID:P05230
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:FGF1|HBGF-1|Beta-endothelial growth factor|ECGF-beta
- Protein/peptide name:FGF-ACIDIC
- Purity:≥95%
- Molecular weight:16 kDa
- Sequence:MFNLPPGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SVGEVYIKST ETGQYLAMDT DGLLYGSQTP NEECLFLERL EENHYNTYIS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, 150 mM sodium sulfate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together