Human Recombinant EGF (from E. coli cells)
Supplier: VWR International
New Product
Certificates
About this item
Epidermal growth factor (EGF) is a growth factor that stimulates the proliferation, differentiation, and survival of epithelial and epidermal cells. EGF contains three intramolecular disulfide bonds and binds in high affinity to the epidermal growth factor receptor (EGFR). EGF is overexpressed in multiple tumor cell lines and promotes resistance to chemotherapy and radiation treatments.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: BALB/c 3T3 cell proliferation; ED50 ≤0.1 ng/ml; Bioactivity ≥1.0×10⁷ units/mg
- Gene ID:P01133
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Urogastrone|URG
- Protein/peptide name:EGF
- Purity:≥95%
- Molecular weight:6.2 kDa
- Sequence:NSDSECPLSH DGYCLHDGVC MYIEALDKYA CNCVVGYIGE RCQYRDLKWW ELR
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together