Human Recombinant ARTEMIN (from E. coli cells)
Supplier: VWR International
New Product
Certificates
About this item
Artemin is a neurotrophin member of the glial cell line-derived neurotrophic factor (GDNF) ligand family. Artemin is highly expressed in the adult pituitary gland, placenta, and trachea, and shows low level expression in the brain, spinal cord, and peripheral tissues. Artemin signals through the RET receptor and GDNF family receptor alpha 3 (GFRα3) co-receptor complex to support neuronal survival.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q5T4W7
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:ARTN|enovin|neublastin
- Protein/peptide name:ARTEMIN
- Purity:≥95%
- Molecular weight:12.0/23.9 kDa
- Sequence:AGGPGSRARA AGARGCRLRS QLVPVRALGL GHRSDELVRF RFCSGSCRRA RSPHDLSLAS LLGAGALRPP PGSRPVSQPC CRPTRYEAVS FMDVNSTWRT VDRLSATACG CLG
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together