Human Recombinant IL-1RA (from E. coli cells)
New Product
Interleukin-1 receptor antagonist (IL-1RA) binds the interleukin 1 receptor (IL-1R) to antagonize interleukin 1 alpha and beta signaling. IL-1RA is secreted by immune cells, epithelial cells, hepatocytes, and adipocytes to inhibit IL-1-induced inflammation.
- High quality
- Low endotoxin
- Affordable
IL-1RA therapy is used to treat auto-inflammatory diseases such as diabetes and rheumatoid arthritis.
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: Inhibition of human IL-1 alpha induced proliferation of D10.G4.1 cells; ED50 ≤50 ng/ml; Bioactivity ≥2.0×10⁴ units/mg
- Gene ID:P18510
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:IL1RN|IL-1F3
- Protein/peptide name:IL-1RA
- Purity:≥95%
- Molecular weight:17.3 kDa
- Sequence:MRPSGRKSSK MQAFRIWDVN QKTFYLRNNQ LVAGYLQGPN VNLEEKIDVV PIEPHALFLG IHGGKMCLSC VKSGDETRLQ LEAVNITDLS ENRKQDKRFA FIRSDSGPTT SFESAACPGW FLCTAMEADQ PVSLTNMPDE GVMVTKFYFQ EDE
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Frequently Bought Together