Human Recombinant IL-8 (from E. coli cells)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 8 (IL-8 or CXCL8) is a member of the CXC cytokine family and is produced by macrophages, epithelial, smooth muscle, and endothelial cells. IL-8 binds the G protein-coupled serpentine receptors CXCR1 and CXCR2. IL-8 recruits innate immune cells, induces phagocytosis, and stimulates angiogenesis.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P10145
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CXCL8|neutrophil chemotactic factor
- Protein/peptide name:IL-8
- Purity:≥95%
- Molecular weight:8.9 kDa
- Sequence:AVLPRSAKEL RCQCIKTYSK PFHPKFIKEL RVIESGPHCA NTEIIVKLSD GRELCLDPKE NWVQRVVEKF LKRAENS
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together