Human Recombinant MIP-3 A (from E. coli cells)
New Product
Macrophage inflammatory protein-3 alpha (MIP-3 α), also called CCL20, is expressed in the liver, lungs, lymph nodes, and peripheral blood lymphocytes. MIP-3 α expression is strongly induced by inflammatory signals, and downregulated by the anti-inflammatory cytokine interleukin 10 (IL-10). MIP-3 α signals through the G protein-coupled receptor CCR6 to function as a chemoattractant to lymphocytes and dendritic cells.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P78556
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CCL20|Exodus-1|LARC
- Protein/peptide name:MIP-3 A
- Purity:≥95%
- Molecular weight:8 kDa
- Sequence:ASNFDCCLGY TDRILHPKFI VGFTRQLANE GCDINAIIFH TKKKLSVCAN PKQTWVKYIV RLLSKKVKNM
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Frequently Bought Together