Order Entry
Canada
Orders LinkContactUsLinkComponent
Human Recombinant MCP-2 (from E. coli cells)

Human Recombinant MCP-2 (from E. coli cells)

Supplier:   VWR International
Human Recombinant MCP-2 (from E. coli cells)
Supplier:  VWR International
Certificates
Human Recombinant MCP-2 (from E. coli cells)

4 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Monocyte chemotactic protein 2 (MCP-2), also known as CCL8, is a cytokine that is important during allergic and inflammatory responses. MCP-2 activates mast cells, eosinophils, and basophils through the G protein-coupled chemokine receptors CCR1, CCR2B, and CCR5. MCP-2 signaling through the CCR5 receptor also functions as a natural inhibitor of the human immunodeficiency virus, type 1 (HIV-1).

  • High quality
  • Low endotoxin
  • Affordable

Caution: For research use only.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 77750-288
  • 77750-290
  • 77750-292
  • 77750-294
  • Protein/peptide name:
  • MCP-2
  • MCP-2
  • MCP-2
  • MCP-2
  • Protein synonyms:
  • CCL8|HC14
  • CCL8|HC14
  • CCL8|HC14
  • CCL8|HC14
  • Protein/peptide type:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Protein Function:
  • Cytokine
  • Cytokine
  • Cytokine
  • Cytokine
  • Species:
  • Human
  • Human
  • Human
  • Human
  • Source:
  • E. coli
  • E. coli
  • E. coli
  • E. coli
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P80075
  • P80075
  • P80075
  • P80075
  • Sequence:
  • QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
  • QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
  • QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
  • QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
  • Protease-free:
  • N
  • N
  • N
  • N
  • Animal-free:
  • N
  • N
  • N
  • N
  • Nuclease-free:
  • N
  • N
  • N
  • N
  • Carrier-free:
  • Y
  • Y
  • Y
  • Y
  • Endotoxin-free:
  • N
  • N
  • N
  • N
  • Endotoxin level:
  • Low
  • Low
  • Low
  • Low
  • Endotoxin content:
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Bioactivity: No biological activity data is available at this time
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
  • ≥95%
  • Grade:
  • RUO
  • RUO
  • RUO
  • RUO
  • Formulation:
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • 0.1% Trifluoroacetic Acid (TFA)
  • Molecular weight:
  • 8.9 kDa
  • 8.9 kDa
  • 8.9 kDa
  • 8.9 kDa
  • Reconstitution instructions:
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Sterile water at 0.1 mg/ml
  • Storage conditions:
  • –20 °C
  • –20 °C
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • RT
  • RT
  • RT
  • RT

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Vial
    100 µg
    Log in for availability
    Log in for availability
    Each
    Each - $647.80
    $647.80
    Each
       $647.80
    Minimum order quantity is 1

    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77750-288
    Protein/peptide name
    MCP-2
    Protein synonyms
    CCL8|HC14
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P80075
    Sequence
    QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8.9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    10 µg
    Log in for availability
    Log in for availability
    Each
    Each - $258.06
    $258.06
    Each
       $258.06
    Minimum order quantity is 1

    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77750-290
    Protein/peptide name
    MCP-2
    Protein synonyms
    CCL8|HC14
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P80075
    Sequence
    QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8.9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    1 mg
    Log in for availability
    Log in for availability
    Each
    Each - $4,645.20
    $4,645.20
    Each
       $4,645.20
    Minimum order quantity is 1

    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77750-292
    Protein/peptide name
    MCP-2
    Protein synonyms
    CCL8|HC14
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P80075
    Sequence
    QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8.9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT
  • Vial
    500 µg
    Log in for availability
    Log in for availability
    Each
    Each - $2,788.70
    $2,788.70
    Each
       $2,788.70
    Minimum order quantity is 1

    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    77750-294
    Protein/peptide name
    MCP-2
    Protein synonyms
    CCL8|HC14
    Protein/peptide type
    Recombinant
    Protein Function
    Cytokine
    Species
    Human
    Source
    E. coli
    Conjugation
    Unconjugated
    Gene ID
    P80075
    Sequence
    QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP
    Protease-free
    N
    Animal-free
    N
    Nuclease-free
    N
    Carrier-free
    Y
    Endotoxin-free
    N
    Endotoxin level
    Low
    Endotoxin content
    Endotoxin LAL (EU/µg) ≤0.1
    Biological activity
    Bioactivity: No biological activity data is available at this time
    Purity
    ≥95%
    Grade
    RUO
    Formulation
    0.1% Trifluoroacetic Acid (TFA)
    Molecular weight
    8.9 kDa
    Reconstitution instructions
    Sterile water at 0.1 mg/ml
    Storage conditions
    –20 °C
    Shipping temperature
    RT