Human Recombinant GM-CSF (from E. coli cells)
Supplier: VWR International
New Product
Certificates
About this item
Granulocyte-macrophage colony-stimulating factor (GM-CSF) is hematopoietic growth factor produced by endothelial cells, monocytes, fibroblasts, and T cells. GM-CSF stimulates the production of neutrophilic granulocytes, macrophages, and mixed granulocyte-macrophage colonies from bone marrow cells.
- High quality
- Low endotoxin
- Affordable
GM-CSF promotes immune system development and regulates neutrophil function during infection. Human and mouse GM-CSF show no cross-reactivity.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: TF-1 cell proliferation; ED50 ≤0.2 ng/ml; Bioactivity ≥5.0×10⁶ units/mg
- Gene ID:P04141
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CSF-2|Pluripoietin-α|MGI-1GM
- Protein/peptide name:GM-CSF
- Purity:≥95%
- Molecular weight:14.6 kDa
- Sequence:MAPARSPSPS TQPWEHVNAI QEARRLLNLS RDTAAEMNET VEVISEMFDL QEPTCLQTRL ELYKQGLRGS LTKLKGPLTM MASHYKQHCP PTPETSCATQ IITFESFKEN LKDFLLVIPF DCWEPVQE
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together