Order Entry
Canada
ContactUsLinkComponent
Human Recombinant GDNF (from E. coli cells)
Human Recombinant GDNF (from E. coli cells)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant GDNF (from E. coli cells)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: C6 proliferation; ED50 ≤3 µg/ml; Bioactivity ≥3.3×10² units/mg
  • Gene ID:
    P39905
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    ATF-1
  • Protein/peptide name:
    GDNF
  • Purity:
    ≥95%
  • Molecular weight:
    15.2/30.4 kDa
  • Sequence:
    MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, 50 mM sodium chloride, pH 8.0
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together