Human Recombinant GDNF (from E. coli cells)
New Product
Glial cell-derived neurotrophic factor (GDNF) is a neurotrophic factor that signals through a multicomponent receptor system to activate receptor tyrosine kinase RET signaling. GDNF promotes dopamine uptake, prevents motor neuron apoptosis, and supports the survival and differentiation of neurons.
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: C6 proliferation; ED50 ≤3 µg/ml; Bioactivity ≥3.3×10² units/mg
- Gene ID:P39905
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:ATF-1
- Protein/peptide name:GDNF
- Purity:≥95%
- Molecular weight:15.2/30.4 kDa
- Sequence:MSPDKQMAVL PRRERNRQAA AANPENSRGK GRRGQRGKNR GCVLTAIHLN VTDLGLGYET KEELIFRYCS GSCDAAETTY DKILKNLSRN RRLVSDKVGQ ACCRPIAFDD DLSFLDDNLV YHILRKHSAK RCGCI
- Endotoxin level:Low
- Formulation:10 mM sodium citrate, 100 mM sodium chloride, pH 4.0
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Frequently Bought Together