Mouse Recombinant MIP-1 B (from E. coli)
New Product
Macrophage inflammatory protein-1 beta (MIP-1 β), also known as CCL4, is produced by macrophages and functions as a mitogen-inducible cytokine. MIP-1 β signals through the chemokine receptor CCR5 to chemoattract immune cells.
- High quality
- Low endotoxin
- Affordable
MIP-1 β induces inflammatory responses, including neutrophil superoxide production. The MIP-1 β and MIP-1 α heterodimer exhibits antiviral activity against the human immunodeficiency virus 1 (HIV-1).
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: THP-1 chemotaxis; ED50 ≤100 ng/ml; Bioactivity ≥NA
- Gene ID:Q5QNV9
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CCL4|ACT-2
- Protein/peptide name:MIP-1 B
- Purity:≥95%
- Molecular weight:7.8 kDa
- Sequence:APMGSDPPTS CCFSYTSRQL HRSFVMDYYE TSSLCSKPAV VFLTKRGRQI CANPSEPWVT EYMSDLELN
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Frequently Bought Together