Mouse Recombinant IFN-G (from E. coli)
New Product
Interferon gamma (IFN-γ) is a type II interferon that is critical during adaptive and innate immune responses to infection. IFN-γ is produced by T cells and natural killer cells following antigen-specific activation.
- High quality
- Low endotoxin
- Affordable
IFN-γ binds IFN-γ receptors (IFN-γ R1 and IFN-γ R2), which are expressed on most immune cells, to activate the JAK-STAT pathway. IFN-γ-induced signaling increases the expression of class 1 major histocompatibility complex (MHC) molecules. Mouse IFN-γ is not cross-reactive with human IFN-γ.
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: Viral CPE assay using EMC virus on L929 cells; ED50 ≤1ng/ml; Bioactivity ≥1.0×10⁶ units/mg
- Gene ID:P01580
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Type II interferon|T cell interferon|Immune Interferon|MAF
- Protein/peptide name:IFN-G
- Purity:≥95%
- Molecular weight:15.7 kDa
- Sequence:MHGTVIESLE SLNNYFNSSG IDVEEKSLFL DIWRNWQKDG DMKILQSQII SFYLRLFEVL KDNQAISNNI SVIESHLITT FFSNSKAKKD AFMSIAKFEV NNPQVQRQAF NELIRVVHQL LPESSLRKRK RSRC
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together