Mouse Recombinant IL-7 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 7 (IL-7) is a hematopoietic cytokine that is an important regulator of B and T cell development. IL-7 is secreted by bone marrow and thymic stromal cells, dendritic cells, intestinal epithelial cells, hepatocytes, and keratinocytes.
- High quality
- Low endotoxin
- Affordable
IL-7 signals through the interleukin 7 receptor (IL-7R) to promote the differentiation of hematopoietic stem cells into T cells, B cells, and natural killer cells. IL-7 is also a regulator of intestinal mucosal lymphocyte proliferation. Human and mouse IL-7 show species cross-reactivity.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: 2E8 cell proliferation; ED50 ≤1 ng/ml; Bioactivity ≥1.0×10⁶ units/mg
- Gene ID:P10168
- Reconstitution instructions:Sterile 10 mM acetic acid at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Lymphopoietin 1 (LP-1)|pre-B cell Factor
- Protein/peptide name:IL-7
- Purity:≥95%
- Molecular weight:15 kDa
- Sequence:MECHIKDKEG KAYESVLMIS IDELDKMTGT DSNCPNNEPN FFRKHVCDDT KEAAFLNRAA RKLKQFLKMN ISEEFNVHLL TVSQGTQTLV NCTSKEEKNV KEQKKNDACF LKRLLREIKT CWNKILKGSI
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together