Human Recombinant GDF-5 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Growth differentiation factor 5 (GDF-5) is a member of the bone morphogenetic protein (BMP) and transforming growth factor beta (TGF-β) families and functions to regulate cell proliferation and differentiation in embryonic and adult tissues.
- High quality
- Low endotoxin
- Affordable
GDF-5 is expressed in the central nervous system and promotes the survival of dopaminergic neurons in animal models of Parkinson's disease. GDF-5 is also important during chondrogenesis and chondrocyte differentiation.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: Alkaline phosphatase activity in ATDC5 cells; ED50 ≤1200 ng/ml; Bioactivity ≥8.3×10² units/mg
- Gene ID:P43026
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CDMP-1
- Protein/peptide name:GDF-5
- Purity:≥95%
- Molecular weight:13.7/27.4 kDa
- Sequence:MAPLATRQGK RPSKNLKARC SRKALHVNFK DMGWDDWIIA PLEYEAFHCE GLCEFPLRSH LEPTNHAVIQ TLMNSMDPES TPPTCCVPTR LSPISILFIDSA NNVVYKQY EDMVVESCGC R
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together