Order Entry
Canada
ContactUsLinkComponent
Human Recombinant OSM (from E. coli)
Human Recombinant OSM (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant OSM (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity assay: TF-1 cell proliferation; ED50 ≤4 ng/ml; Bioactivity ≥2.5×10⁵ units/mg
  • Gene ID:
    P13725
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    Onc-M
  • Protein/peptide name:
    OSM
  • Purity:
    ≥95%
  • Molecular weight:
    23.8 kDa
  • Sequence:
    MAAIGSCSKE YRVLLGQLQK QTDLMQDTSR LLDPYIRIQG LDVPKLREHC RERPGAFPSE ETLRGLGRRG FLQTLNATLGCVLHRLADLE QRLPKAQDLE RSGLNIEDLE KLQMARPNIL GLRNNIYCMA QLLDNSDTAE PTKAGRGASQ PPTPTPASDA FQRKLEGCRFLHGYHRFMHS VGRVFSKWGE SPNRSRRHSP HQALRKGVRR
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together