Human Recombinant GALECTIN-3 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Galectin-3 belongs to the lectin family of carbohydrate binding proteins. Galectin-3 is expressed by a wide range of cell types including activated T cells, tumor cells, macrophages, osteoclasts, fibroblasts, and epithelial cells. Galectin-3 has specific binding affinity for beta-galactoside sugar moieties and has functional roles during development, innate immunity, cell apoptosis, and tumor metastasis.
- High quality
- Low endotoxin
- Affordable
Galectin-3 is associated with cancer, heart failure, stroke, and inflammation. The amino acid sequences of human and mouse Galectin-3 proteins share 80% homology.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P17931
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Galactose-specific lectin-3|IgE-binding protein|MAC2|L-29|CPB-35
- Protein/peptide name:GALECTIN-3
- Purity:≥95%
- Molecular weight:26.2 kDa
- Sequence:MADNFSLHDA LSGSGNPNPQ GWPGAWGNQP AGAGGYPGAS YPGAYPGQAP PGAYPGQAPP GAYPGAPGAY PGAPAPGVYP GPPSGPGAYP SSGQPSATGA YPATGPYGAP AGPLIVPYNL PLPGGVVPRM LITILGTVKP NANRIALDFQ RGNDVAFHFN PRFNENNRRV IVCNTKLDNN WGREERQSVF PFESGKPFKI QVLVEPDHFK VAVNDAHLLQ YNHRVKKLNE ISKLGISGDI DLTSASYTMI
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together