Human Recombinant IL-16 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 16 (IL-16) is produced by CD4+ and CD8+ T cells and functions as a chemoattractant for lymphocytes, monocytes, eosinophils, dendritic cells, and Langerhans cells. Additionally, IL-16 upregulates the expression of the interleukin 2 receptor (CD25), modulates T lymphocyte growth, and suppresses the replication of the human immunodeficiency virus (HIV) and simian immunodeficiency viruses (SIV).
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q14005
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Lymphocyte Chemoattractant Factor (LCF)
- Protein/peptide name:IL-16
- Purity:≥95%
- Molecular weight:13.4 kDa
- Sequence:MPDLNSSTDS AASASAASDV SVESTAEATV CTVTLEKMSA GLGFSLEGGK GSLHGDKPLT INRIFKGAAS EQSETVQPGD EILQLGGTAM QGLTRFEAWN IIKALPDGPV TIVIRRKSLQ SKETTAAGDS
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together