Order Entry
Canada
ContactUsLinkComponent
Human Recombinant IGF-II (from E. coli)
Human Recombinant IGF-II (from E. coli)
Supplier:  VWR International
New Product
Certificates
undefined
Human Recombinant IGF-II (from E. coli)
Supplier:  VWR International
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: rh IGF-II activity is tested in human breast cancer cell line MCF-7; ED50 ≤6 ng/ml; Bioactivity ≥1.7×10⁵ units/mg
  • Gene ID:
    P01344
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    Somatomedin A
  • Protein/peptide name:
    IGF-II
  • Purity:
    ≥95%
  • Molecular weight:
    7.5 kDa
  • Sequence:
    AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

Specifications

Frequently Bought Together