Mouse Recombinant TH9 CELL POLARIZING PB (from E. coli)
Catalog # 77753-622
Supplier: VWR International
New Product
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:Each
- Storage conditions:At or below –20 °C
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:TH9 CELL POLARIZING PB
- Endotoxin level:Low
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
About this item
Cytokines for polarizing mouse T cells into Th9 cells. Th9 cells are a unique subset of T helper cells that produce IL-9. Although the exact function of IL-9 producing T cells is yet to be completely clear it is proposed that these cells could be important in inflammation and helminth infections. Th9 cells can be generated from naive CD4⁺ T cells in culture by culturing them in the presence of IL-4 and TGF-β 1. It also appears that IFNγ and IL-27 can inhibit the generation of Th9 cells.
High quality
- Low endotoxin
- Affordable
Caution: For research use only.