Human/Mouse Recombinant TGF-B 3 (from E. coli)
New Product
Transforming growth factors (TGFs) are multifunctional peptides that regulate growth and differentiation in most cell types. The TGF-β family of proteins signal through serine/threonine kinase receptors. TGF-β isoforms (TGF-β1, -β2, and –β3) have overlapping, yet distinct biological actions in developing and adult tissues. TGF-β3 is an important factor in regulating cell adhesion and accelerating wound repair. TGF-β3 also functions during osteoblast proliferation, chemotaxis, and collagen synthesis.
High quality
- Low endotoxin
- Affordable
: For research use only.
- Source:
- Species:
- Environmentally Preferable:
Frequently Bought Together