Order Entry
Canada
ContactUsLinkComponent
 
Human/Mouse Recombinant TGF-B 3 (from E. coli)
New Product
 
undefined
Human/Mouse Recombinant TGF-B 3 (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
    Mouse
  • Storage conditions:
    At or below –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation; ED50 ≤1 ng/ml; Bioactivity ≥1.0×10⁶ units/mg
  • Gene ID:
    P10600/P17125
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein/peptide name:
    TGF-B 3
  • Purity:
    ≥95%
  • Molecular weight:
    12.9/25.7 kDa
  • Sequence:
    MALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCS
  • Endotoxin level:
    Low
  • Formulation:
    0.1% Trifluoroacetic acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together