Horse Recombinant PDGF-BB (from E. coli)
New Product
Platelet-derived growth factor (PDGF) is an important regulator of cell growth, proliferation, and angiogenesis. PDGF synthesis is induced by IL-1, IL-6, TNF-α, TGF-β, and EGF signaling. PDGF functions as a mitogenic growth hormone on cells of mesenchymal lineage, such as smooth muscle and glial cells. PDGF is also stored in the alpha-granules of platelets and is released upon adherence to traumatized tissues. PDGF is a dimeric glycoprotein formed by two A chains (AA), two B chains (BB), or as a heterodimer with an A and a B chain (AB). The PDGF dimer binds the cell surface receptor tyrosine kinases PDGFR-α and PDGFR-β.
High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Horse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: NR6R 3T3 proliferation; ED50 ≤30 ng/ml; Bioactivity ≥3.3×10⁴ units/mg
- Gene ID:XP_005606703
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:GDGF|ODGF
- Protein/peptide name:PDGF-BB
- Purity:≥95%
- Molecular weight:24.8 kDa
- Sequence:MSLGSLAVAE PAMIAECKTR TEVFEISRRL IDRTNANFLV WPPCVEVQRC SGCCNNRHVQ CRPTQVQLRP VQVRKIEIVR KKPTFKKATV TLEDHLACKC ETVGAARPVT
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together