Mouse Recombinant TPO (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Thrombopoietin (TPO) is a growth factor that is produced by liver and kidney tissues. TPO binds the TPO receptor (CD110) to promote megakaryocyte maturation, differentiation, and the production of platelets.
High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity assay: MO7e cell proliferation; ED50 ≤5 ng/ml; Bioactivity ≥2.0×10⁵ units/mg
- Gene ID:P40226
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Megakaryocyte Colony Stimulating Factor|c-MPL Ligand|MGDF
- Protein/peptide name:TPO
- Purity:≥95%
- Molecular weight:18.8 kDa
- Sequence:MSPVAPACDP RLLNKLLRDS HLLHSRLSQC PDVDPLSIPV LLPAVDFSLG EWKTQTEQSK AQDILGAVSL LLEGVMAARG QLEPSCLSSL LGQLSGQVRL LLGALQGLLG TQLPLQGRTT AHKDPNALFL SLQQLLRGKV RFLLLVEGPT LCVRRTLPTT AVPSSTSQLL TLNKF
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together