Mouse Recombinant B-NGF (from E. coli)
New Product
Nerve growth factor beta (β-NGF) is a neurotrophic factor that is important for the development and maintenance of sensory and sympathetic neurons. β-NGF signals through the low affinity nerve growth factor receptor (LNGFR) and the tropomyosin receptor kinase A (TrkA) to activate PI3K, Ras, and PLC signaling pathways. β-NGF is also involved in the growth, differentiation, and survival of B lymphocytes. Human, mouse, and rat β-NGF proteins are cross-reactive.
High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: TF-1 cell proliferation; ED50 ≤5 ng/ml; Bioactivity ≥2.0×10⁵ units/mg
- Gene ID:P01139
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:beta-NGF
- Protein/peptide name:B-NGF
- Purity:≥95%
- Molecular weight:13.6/27.2 kDa
- Sequence:MSSTHPVFHM GEFSVCDSVS VWVGDKTTAT DIKGKEVTVL AEVNINNSVF RQYFFETKCR ASNPVESGCR GIDSKHWNSY CTTTHTFVKA LTTDEKQAAW RFIRIDTACV CVLSRKATRR G
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together