Human Recombinant IL-17E (from E. coli)
New Product
Interleukin 17E (IL-17E), also known as IL-25, is a member of the IL-17 family of cytokines. IL-17E binds to the IL-17RB receptor to stimulate the secretion of the proinflammatory interleukin 8 (IL-8) chemokine and to induce the activation of nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB).
High quality
- Low endotoxin
- Affordable
: For research use only.
- Source:
- Species:
- Environmentally Preferable:
Frequently Bought Together