Autoclavable Tyvek Breather Bags, Easy Chevron Peel Feature and Heat Seal Closure, Steam Indicator
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
New Product
Certificates
About this item
A significant upgrade over paper-based steam sterilization pouches. These Tyvek® peel pouches will not rip or puncture like paper-based autoclave pouches routinely do.
- Manufactured and packaged in an ISO certified cleanroom Chevron peel opening Autoclavable at 121 to 125 °C Tear and puncture resistant Hydrophobic Lint-free
The medical Grade 1073B Tyvek® material has the highest microbial barrier properties of any steam sterilization compatible materials. These pouches are also compatible with VHP Decontamination cycles commonly used with Isolator technology. Will not absorb or adsorb water.
Closure type: Heat seal
Opening: Peel
Delivery:
Supplied with Certificate of Conformance.
Frequently Bought Together