Order Entry
Canada
ContactUsLinkComponent
 
Pig Recombinant IL-4 (from E. coli)
New Product
 
undefined
Pig Recombinant IL-4 (from E. coli)
New Product

 

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Pig
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤0.1
  • Biological activity:
    Bioactivity assay: TF-1 cell proliferation; ED50 ≤2 ng/ml; Bioactivity ≥5.0×10⁵ units/mg
  • Gene ID:
    Q04745
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    Y
  • Protein synonyms:
    B-cell stimulatory factor-1|BSF-1|BCDF|BCGF
  • Protein/peptide name:
    IL-4
  • Purity:
    ≥95%
  • Molecular weight:
    12.7 kDa
  • Sequence:
    MHKCDITLQE IIKTLNILTA RKNSCMELPV TDVFAAPENT TEKETFCRAS TVLRHIYRHH TCMKSLLSGL DRNLSSMANM TCSVHEAKKS TLKDFLERLK TIMKEKYSKC
  • Endotoxin level:
    Low
  • Formulation:
    10 mM sodium phosphate, pH 7.5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together