Rat Recombinant IL-17A (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 17A (IL-17A), also known as CTLA-8, is a member of the IL-17 family of proteins. IL-17A is a proinflammatory cytokine that is secreted by activated CD4+ and CD8+ T lymphocytes. IL-17A acts through its receptor, IL-17R, to promote increased cytokine and chemokine secretion. In turn, the cytokines and chemokines mediate the immunoregulatory function of IL-17A by promoting the proliferation, maturation, and chemoattraction of neutrophils to inflammatory sites. Elevated levels of IL-17A are associated with rheumatoid arthritis, airway inflammation, allograft rejection, inflammatory bowel disease, psoriasis, cancer, and multiple sclerosis. Human, mouse, and rat IL-17A show activity on mouse cells.
High quality
Low endotoxin
Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q61453
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:IL-17|CTLA-8
- Protein/peptide name:IL-17A
- Purity:≥95%
- Molecular weight:15.1/30.3 kDa
- Sequence:MAVLIPQSSV CPNAEANNFL QNVKVNLKVL NSLSSKASSR RPSDYLNRST SPWTLSRNED PDRYPSVIWE AQCRHQRCVN AEGKLDHHMN SVLIQQEILV LKREPEKCPF TFRVEKMLVG VGCTCVSSIV RHAS
- Endotoxin level:Low
- Formulation:10 mM sodium citrate, pH 3.0
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together