Mouse Recombinant IL-1A (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Interleukin 1 alpha (IL-1α) is expressed by epithelial cells, activated macrophages, neutrophils, and endothelial cells to regulate immune responses. IL-1α signals through the IL-1 receptor, type 1 (IL-1R1) to activate the myeloid differentiation primary response 88 (MYD88) signaling pathway, which contains the cytoplasmic toll/IL-1 receptor (TIR) domain adapter. IL-1α and the independently regulated IL-1β protein have overlapping proinflammatory activities to induce adhesion molecule expression on epithelial cells, control fever induction, initiate rheumatoid arthritis, and promote septic shock.
High quality
Low endotoxin
Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: D10.G4.1 proliferation; ED50 ≤0.01 ng/ml; Bioactivity ≥1.0×10⁸ units/mg
- Gene ID:P01582
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein/peptide name:IL-1A
- Purity:≥95%
- Molecular weight:18.1 kDa
- Sequence:MSAPYTYQSD LRYKLMKLVR QKFVMNDSLN QTIYQDVDKH YLSTTWLNDL QQEVKFDMYA YSSGGDDSKY PVTLKISDSQ LFVSAQGEDQ PVLLKELPET PKLITGSETD LIFFWKSINS KNYFTSAAYP ELFIATKEQS RVHLARGLPS MTDFQIS
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together