Mouse Recombinant FLT-3 LIGAND (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Fms-related tyrosine kinase 3 ligand (FLT-3 ligand) is a growth factor that regulates hematopoietic cell proliferation. FLT-3 ligand signaling is transmitted through the fms-related tyrosine kinase 3 (FLT-3) receptor. FLT-3 ligand promotes the long-term expansion and differentiation of pro-B cells in the presence of interleukin 7 (IL-7) or in combination of IL-7 and interleukin 3 (IL-3).
High quality
Low endotoxin
Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: OCI-Aml5 cell proliferation; ED50 ≤10 ng/ml; Bioactivity ≥1.0×10⁵ units/mg
- Gene ID:P49772
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:Flt3L|Fms-related tyrosine kinase 3 ligand
- Protein/peptide name:FLT-3 LIGAND
- Purity:≥95%
- Molecular weight:18.6 kDa
- Sequence:MTPDCYFSHS PISSNFKVKF RELTDHLLKD YPVTVAVNLQ DEKHCKALWS LFLAQRWIEQ LKTVAGSKMQ TLLEDVNTEI HFVTSCTFQP LPECLRFVQT NISHLLKDTC TQLLALKPCI GKACQNFSRC LEVQCQPDSS TLLPPRSPIA LEATELPEPR PRQ
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together