Human Recombinant LIF (from E. coli)
New Product
Leukemia inhibitory factor (LIF) is a member of the interleukin 6 (IL-6) family that is made by a variety of adult and embryonic tissues. LIF signals through the glycoprotein 130 (gp130)/LIF receptor (LIFR) heterodimer to activate STAT3 and MAPK signaling. LIF functions during hematopoietic differentiation, neuronal cell differentiation, kidney development, and inflammatory processes. Human LIF may also be an important factor during human embryonic stem cell (hESC) self-renewal, pluripotency, and embryonic implantation.
High quality
Low endotoxin
Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: TF-1 cell proliferation; ED50 ≤0.2 ng/ml; Bioactivity ≥5.0×10⁶ units/mg
- Gene ID:P15018
- Reconstitution instructions:Sterile 10 mM acetic acid at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Leukocyte Inhibitory Factor
- Protein/peptide name:LIF
- Purity:≥95%
- Molecular weight:19.8 kDa
- Sequence:MSPLPITPVN ATCAIRHPCH NNLMNQIRSQ LAQLNGSANA LFILYYTAQG EPFPNNLDKL CGPNVTDFPP FHANGTEKAK LVELYRIVVY LGTSLGNITR DQKILNPSAL SLHSKLNATA DILRGLLSNV LCRLCSKYHV GHVDVTYGPD TSGKDVFQKK KLGCQLLGKY KQIIAVLAQA F
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together