Human/Mouse Recombinant IHH (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Indian hedgehog (IHH) is an essential signaling factor that is secreted in the gut, cartilage, and bone during embryonic development. IHH acts through the patched (PTC) receptor to induce transcriptional changes important for bone and cartilage development. IHH also induces the expression of parathyroid hormone-related peptide (PTHrP), which in turn mediates IHH activity during chondrocyte differentiation, forming a negative feedback loop.
- High quality Low endotoxin Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:HumanMouse
- Storage conditions:At or below –20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q14623/P97812
- Reconstitution instructions:10 mM HCl at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:HHG-2
- Protein/peptide name:IHH
- Purity:≥95%
- Molecular weight:19.9 kDa
- Sequence:MIIGPGRVVG SRRRPPRKLV PLAYKQFSPN VPEKTLGASG RYEGKIARSS ERFKELTPNY NPDIIFKDEE NTGADRLMTQRCKDRLNSLA ISVMNQWPGV KLRVTEGWDE DGHHSEESLH YEGRAVDITT SDRDRNKYGL LARLAVEAGF DWVYYESKAH VHCSVKSEHSAAAKTGG
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together