Pig Recombinant IL-8 (from E. coli)
New Product
Interleukin 8 (IL-8 or CXCL8) is a member of the CXC cytokine family and is produced by macrophages, epithelial, smooth muscle, and endothelial cells. IL-8 binds the G protein-coupled serpentine receptors CXCR1 and CXCR2. IL-8 recruits innate immune cells, induces phagocytosis, and stimulates angiogenesis.
- High quality Low endotoxin Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Pig
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P26894
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:Y
- Protein synonyms:CXCL8|neutrophil chemotactic factor
- Protein/peptide name:IL-8
- Purity:≥95%
- Molecular weight:9.1 kDa
- Sequence:ARVSAELRCQ CINTHSTPFH PKFIKELRVI ESGPHCENSE IIVKLVNGKE VCLDPKEKWV QKVVQIFLKR TEKQQQQQ
- Endotoxin level:Low
- Formulation:5 mM sodium phosphate, 20 mM sodium chloride, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Frequently Bought Together