Human Recombinant MCP-4 (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Monocyte chemotactic protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as interleukin 1 (IL-1) and tumor necrosis factor alpha (TNF-α). MCP-4 is a ligand for the G protein coupled chemokine receptors CCR2, CCR3, and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils, and basophils during inflammation and allergic responses.
High quality
Low endotoxin
Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:Q99616
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:CCL13|NCC-1
- Protein/peptide name:MCP-4
- Purity:≥95%
- Molecular weight:8.6 kDa
- Sequence:QPDALNVPST CCFTFSSKKI SLQRLKSYVI TTSRCPQKAV IFRTKLGKEI CADPKEKWVQ NYMKHLGRKA HTLKT
- Endotoxin level:Low
- Formulation:0.1% Trifluoroacetic Acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together