Rat Recombinant M-CSF (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Macrophage colony stimulating factor (M-CSF) is a hematopoietic growth factor that is widely produced by a variety of cells. M-CSF stimulates the proliferation and differentiation of hematopoietic stem cells into monocyte and macrophage cell types. M-CSF also acts through the colony stimulating factor 1 receptor (CSF1R) to modulate processes involved in immunology, bone metabolism, fertility, and pregnancy.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: NFS-60 Proliferation; ED50 ≤10 ng/ml; Bioactivity ≥1.0×10⁵ units/mg
- Gene ID:Q8JZQ0
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:MGI-IM|CSF-1
- Protein/peptide name:M-CSF
- Purity:≥95%
- Molecular weight:18.1/36.2 kDa
- Sequence:MEVSEHCSHM IGNGHLQILQ QLIDSQMETA CLIEYKFVDQ EQLDDPVCYL KKAFVLVQVI IEETMRFKDN TPNANATERL QELSMKLNSC FIKDYKEQNE ACVQTYKESP LRLLEKIKNF FNETKNFLEK DWNIFSKNCN DSLAKCSSRD VVTKP
- Endotoxin level:Low
- Formulation:10 mM sodium phosphate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together