Mouse Recombinant IGF-II (from E. coli)
Supplier: VWR International
New Product
Certificates
About this item
Insulin-like growth factor II (IGF-II) is an important fetal growth hormone made by theca cells during gestation. IGF-II engages the IGF-I receptor (IGF1R) to mediate embryonic growth. IGF-II also binds the sink IGF-II receptor (IGF2R) leading to IGF-II degradation.
- High quality
- Low endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0.1
- Biological activity:Bioactivity assay: FDC-P1 cell proliferation; ED50 ≤50 ng/ml; Bioactivity ≥2.0×10⁴ units/mg
- Gene ID:P09535
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:Somatomedin A
- Protein/peptide name:IGF-II
- Purity:≥95%
- Molecular weight:7.4 kDa
- Sequence:AYGPGETLCG GELVDTLQFV CSDRGFYFSR PSSRANRRSR GIVEECCFRS CDLALLETYC ATPAKSE
- Endotoxin level:Low
- Formulation:0.1% trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Specifications
Frequently Bought Together